Key Documents
Z221767
Filter-regulator-lubricator
gauge 1 1/2 in., pressure 0-160 psi, fitting NPTM 1/4 in.
About This Item
UNSPSC Code:
41103801
NACRES:
NB.72
parameter
0-160 psi pressure
gauge
1 1/2 in.
General description
Filters water aerosols and foreign particles larger than 40μm from supply air, then automatically lubricates air going through motor. Oil feed rate should be adjusted to 1drop every 5minutes. Complete assembly as shown.
Application
For use with air-powered stirrers and motors.
Choose from one of the most recent versions:
Certificates of Analysis (COA)