Key Documents
Z220531
Magic Touch ice bucket
black
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
manufacturer/tradename
Bel-Art M18848-4002
parameter
−321-200 °F temperature
capacity
1 gal (4 L)
top diam.
10 1/4 in. (27 cm)
color
black
General description
Expanded PVC bucket is light-weight, leak-proof, and unbreakable. Excellent insulative properties make them ideal for containing liquid nitrogen, dry ice/acetone, and ice/saline. One piece with lid.
Legal Information
Magic Touch is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)