Key Documents
Z217956
Supelco
Feed Tube Adapters
size 25 mm
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
manufacturer/tradename
Ace Glass 582628
technique(s)
LPLC: suitable
size
25 mm
General description
Ace-Thred #7 at top with nylon bushing and FETFE O-ring for use with feed tubes listed. Size refers to I.D. of tubing above lower thread and corresponds to M-M column diameter
Choose from one of the most recent versions:
Certificates of Analysis (COA)