Key Documents
Z217859
Supelco
Feed Tube
L 76 mm, with hose connection
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
manufacturer/tradename
Ace Glass 583104
technique(s)
LPLC: suitable
L
76 mm
General description
Permits packing of column without excessive drop impact. Also for introduction of effluent or for pressurized operation. Use with feed tube adapters.
Choose from one of the most recent versions:
Certificates of Analysis (COA)