Key Documents
Z215538
Rigid PTFE sleeve
joint: ST/NS 34/35
Synonym(s):
ptfe sleeves
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
joint
joint: ST/NS34/35
manufacturer/tradename
Cowie
General description
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)