High resolution vapor-phase FT-IR spectral library for use with Spectral ID®, GRAMS®, and other spectral search software. The library contains spectra at 4 cm-1 resolution by 16-bit ordinate precision, which are suitable for doing searches using spectral subtraction
More than 5,000 FT-IR spectra of organics produced using high resolution gas chromatography
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
GRAMS is a registered trademark of Thermo Electron Corp.
Spectral ID is a registered trademark of Galactic Industries Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)