Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z213381

Sigma-Aldrich

Carton sealing tape dispenser

hand-held pull and tear method

Synonym(s):

hand held dispenser, sealing tape, tape dispensers, tape gun

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.35

sterility

non-sterile

manufacturer/tradename

Sigma-Aldrich

Application

Use with product: Z213373

The dispenser allows for a convenient length before cut-off and helps the operator apply (can help smooth down) the tape. It allows the tape to be taken to the point of application for operator ease and then cuts tape length.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service