Key Documents
Z206989
Nylon inspection gloves
size M
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
Nylon
white gloves
size
M
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pkg of 12 pair
manufacturer/tradename
Sigma-Aldrich
General description
Thin tricot knit conforms to hand, is lint-free, and provides excellent touch sensitivity. Use for inspection or as a glove liner. Ambidextrous.
Choose from one of the most recent versions:
Certificates of Analysis (COA)