material
polyethylene
sterility
non-sterile
feature
graduations: no
packaging
pack of 5
manufacturer/tradename
Bel-Art
L
39 cm (15 1/4 in.)
bulb draw
20 mL
General description
One-piece molded polyethylene sampler with bellows and calibrated sampler tube to indicate collected volume. Long tapered tips have a 4 mm opening.
Choose from one of the most recent versions:
Certificates of Analysis (COA)