Key Documents
Z205818
Sigma-Aldrich
Hazardous accumulation labels
4 × 6 in. Roll of 500
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
material
paper
packaging
pkg of 1 roll (500 labels)
manufacturer/tradename
Sigma-Aldrich
L × W
10.1 cm × 15.25 cm
General description
Comply with EPA regulations for hazardous waste container markings. Fill in the accumulation start date and name(s) of the hazardous materials in the space provided and apply label to container. For indoor use only. 4 × 6 in. Roll of 500.
Choose from one of the most recent versions:
Certificates of Analysis (COA)