Key Documents
Z205095
Sigma-Aldrich
Laboratory shelf can
capacity 5 gal
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
material
terneplated steel
manufacturer/tradename
Justrite 10807
availability
not available in EU
H × diam.
11 1/2 in. × 17 in.
capacity
5 gal
General description
Safety can with self-closing brass faucet for controlled dispensing of hazardous liquids. Faucet is mounted two inches above bottom. FM approved.
Choose from one of the most recent versions:
Certificates of Analysis (COA)