Key Documents
Z199826
Sigma-Aldrich
Oberon chemically-resistant safety goggles
Indirect vent
About This Item
UNSPSC Code:
46181802
material
clear vinyl frame
polycarbonate lens
availability
not available in EU
lens thickness
0.050 in.
General description
Resist mild acids, caustics, aromatics, hydrocarbons, and methylene chloride (excludes hydrofluoric acid). Coated with chemically resistant Silicote-6. Meets ANSI Z87.1-1989 requirements.
Choose from one of the most recent versions:
Certificates of Analysis (COA)