Key Documents
Z198765
Sigma-Aldrich
NFPA labels
size 4 in. × 4 in.
About This Item
UNSPSC Code:
55121611
material
paper
packaging
pkg of 250
manufacturer/tradename
Sigma-Aldrich
size
4 in. × 4 in.
General description
Identify the degree of flammability, health, reactivity, and special hazards. Colored diamond areas can be numbered in the NFPA 704 Hazard Rating System from 0 to 4. Pressure-sensitive paper.
Choose from one of the most recent versions:
Certificates of Analysis (COA)