Key Documents
Z198749
Sigma-Aldrich
Write-on hazardous-communication labels
size 4 in. × 4 in.
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
material
paper
packaging
pkg of 250
manufacturer/tradename
Sigma-Aldrich
size
4 in. × 4 in.
General description
Used to identify chemical names; hazard ratings from 0 to 4 for flammability, health, and reactivity; and personal protective equipment. Pressure-sensitive paper.
Legal Information
Write-on is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)