Key Documents
Z197750
Sigma-Aldrich
Dual-action drum vent
valve opens at 5 psi, 1ea
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
manufacturer/tradename
Justrite 08005
General description
Automatic pressure relief valve opens at 5 psi internal pressure. Vacuum relief is automatic and permits 4 gpm flow through a standard 3/4 in. faucet. HDPE with steel flange.
Choose from one of the most recent versions:
Certificates of Analysis (COA)