Z194441
PE safety-bottle jacket
fits 4 L and 1 gal bottles
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
feature
Holds 1x 4 L -1 gal bottle
manufacturer/tradename
Justrite 12009
availability
not available in EU
General description
Designed to fit 4 L and 1 gal bottles. Lab bottles fit into the bottom half of the cover and lid twists securely for safe pouring. Jacket also acts as a shock absorber, minimizing breakage and spills due to bumps or falls.
Choose from one of the most recent versions:
Certificates of Analysis (COA)