Key Documents
Z192465
Sigma-Aldrich
BD Precisionglide® syringe needles
Synonym(s):
cannula
About This Item
UNSPSC Code:
12352200
Pricing and availability is not currently available.
packaging
pkg of 100 ea
manufacturer/tradename
Becton-Dickinson & Co.
General description
0.75 in. length, 22 gauge
  • Fits any Luer lock, slip tip, or eccentric tip syringe
  • SS Microlance regular-bevel point
  • Siliconized to reduce resistance
  • PP Luer hub color coded to needle gauge
  • Packaged as individual fiber-free, peel-apart packages in strips of five sterile needles
  • A sheath protects point and cannula
  • All packages carry lot control numbers to insure sterility
  • NoKor needles have scalpel like tip
Legal Information
Precisionglide is a registered trademark of Becton-Dickinson & Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)