Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z192414

Sigma-Aldrich

BD Precisionglide® syringe needles

gauge 25, L 1 1/2 in.

Synonym(s):

cannula

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122002
NACRES:
NB.03

packaging

pkg of 100 ea

manufacturer/tradename

Becton-Dickinson & Co. 305127

L

1 1/2 in.

gauge

25

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

  • Fits any Luer lock, slip tip, or eccentric tip syringe
  • SS Microlance regular-bevel point
  • Siliconized to reduce resistance
  • PP Luer hub color coded to needle gauge
  • Packaged as individual fiber-free, peel-apart packages in strips of five sterile needles
  • A sheath protects point and cannula
  • All packages carry lot control numbers to insure sterility
  • NoKor needles have scalpel like tip

Legal Information

Precisionglide is a registered trademark of Becton-Dickinson & Co.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service