Key Documents
Z188050
Filter-paper support discs
diam. 90 mm
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
material
high-density polyethylene membrane
sterility
non-sterile
diam.
90 mm
General description
Made from high density polyethylene, these discs have excellent chemical resistance. They are useful for holding up filter paper. In this way the blocking of the holes is avoided and the filtering surface of the funnel is free. Will withstand temperatures up to 110°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)