Key Documents
Z186813
Sigma-Aldrich
All-purpose sampling containers
PE lids for 41/2 & 8oz sampling containers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
General description
Durable and translucent PP containers resist acids, bases, most organic solvents, and common laboratory reagents. Internally molded dual graduations in increments of 1 oz and 20 mL. Economical enough to discard after one use, or strong enough to reuse. Tight-fitting PE lid not included. Nonsterile.
Other Notes
Supplied without lid. No lid available.
Choose from one of the most recent versions:
Certificates of Analysis (COA)