Key Documents
Z185140
Sigma-Aldrich
Aluminum foil
W × L 12 in. × 1,000 ft, thickness 0.0007 in.
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
sterility
non-sterile
manufacturer/tradename
Sigma-Aldrich
W × L
12 in. × 1,000 ft
thickness
0.0007 in.
General description
Easily covers laboratory dishes and forms inexpensive liners for bench tops, drawers, and trays. Easy cleanup in case of spill or contamination.
Application
Aluminum foil has been used:
  • to cover the culture plates
  • to store coverslips for months
  • to cover tubes and bottle-filling system for autoclaving
Choose from one of the most recent versions:
Certificates of Analysis (COA)