Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z183679

Poly-net® protective netting

red, diam. 2-4 in., Close-mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

diam.

2-4 in.

thickness

0.07-1.2 mm , approximate

color

red

Looking for similar products? Visit Product Comparison Guide

General description

PE net sleeve stretches around glassware to prevent scratching,chipping or breaking. Netting captures flying glass if container shatters. Easily cut to any length. Color-coded for size identification.

Application

Funnels, flasks, and condensers

Legal Information

Poly-net is a registered trademark of Norddeutsche Seekabelwerke GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service