MilliporeSigma
All Photos(4)
Key Documents
Z181706
Supelco
Bottom-drip adapter for large columns
Luer tip and flow-regulator valve, 0 to 1mm adjustable bore, Nylon
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
material
Nylon
manufacturer/tradename
Ace Glass 583723
technique(s)
LPLC: suitable
General description
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred #50 and include one FETFE® O-ring.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)