Key Documents
Z180319
Supelco
Tubing connector for large columns
Nut, tube I.D. 1.5 mm
About This Item
UNSPSC Code:
41121813
NACRES:
SB.52
manufacturer/tradename
Ace Glass 585508
technique(s)
LPLC: suitable
tube I.D.
1.5 mm
General description
TFE nut with 1/4-28 UNF 2B male thread connects small I.D. tubing to bottom drip adapter. KEL-F insert mechanically wedges tubing against fitting. For vacuum or pressure applications. Tubing-insertion tool required for assembly. Order nut, insert, and insertion tool separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)