Key Documents
Z179949
Supelco
Injection-port adapter
Injection port cap
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
manufacturer/tradename
Ace Glass 580702
technique(s)
LPLC: suitable
General description
PTFE; allows direct on-column injection at the center of filter disc at top of column without stopping solvent flow. Top thread is 5/16 -24 UNF for use with injection-port cap that holds septum. Side female thread is 1/4 -28 UNF to accommodate tubing adapters or other miniature plumbing systems. Bottom male thread refers to Ace-Thred number on columns. Complete item is supplied with one PTFE-coated silicone-rubber septum. Not suitable for larger columns. Order paper filter discs separately with column end-fitting adapters.
fits adapters with #7 and 11 Ace-Threds
Choose from one of the most recent versions:
Certificates of Analysis (COA)