Key Documents
Z179841
Supelco
Michel-Miller column coupling
Ace-Thred for #7-7
About This Item
UNSPSC Code:
41115710
NACRES:
SB.52
manufacturer/tradename
Ace Glass 580503
technique(s)
LPLC: suitable
fitting
Ace-Thred for #7-7
General description
PTFE with 316 SS Liner connects M-M columns when packing. Makes a leaktight seal without O-rings. Male Ace Thred matches thread size of columns.
Choose from one of the most recent versions:
Certificates of Analysis (COA)