Key Documents
Z179647
Supelco
Michel-Miller end-fitting adapter
Ace-Thred for #25
About This Item
UNSPSC Code:
23151817
NACRES:
SB.52
manufacturer/tradename
Ace Glass 580122
technique(s)
LPLC: suitable
fitting
Ace-Thred for #25
General description
Use at either end of column. 1/4 -28 female thread at top accepts standard miniature plumbing systems. Male Ace-Thred matches thread size of M-M columns. Bore is 1.5 mm. Use with filter discs to retain column-packing material. Order adapter and filter discs separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)