Key Documents
Z179612
Supelco
Michel-Miller safety shield
size G
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
size
G
manufacturer/tradename
Ace Glass 579864
technique(s)
LPLC: suitable
General description
Ensures safe operation of M-M columns at pressures to 300 psi. Top thread of column is hung between two-piece aluminum cap; cap is then held by plastic shield. Clamp shield, not column. The 3 mm acrylic wall is not resistant to most solvents.
Choose from one of the most recent versions:
Certificates of Analysis (COA)