Key Documents
Z174424
Sigma-Aldrich
Glass water bath
O.D. × I.D. 115 mm × 105 mm, fits up to 400 mL tall form beaker or flask
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
feature
fits up to 400 mL tall form beaker or flask
O.D. × I.D.
115 mm × 105 mm
General description
Accommodates up to 400 mL tall form flask or beaker. Fits on magnetic stirrer. Reaction in flask or beaker can be readily observed.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1