With two 1/4 in. NPTF end valve threads. Can be used to store or transport liquids and gases at pressures up to 1,800 psig at room temperature; also, as pressure purge accumulator, reaction vessel, gas sampling for chromatography, lecture bottle for lab use, and for condensate sampling and transporting in fossil fuel and nuclear power plants.
Hot-spun (not welded) from seamless, 304 grade SS tubing for maximum integrity. Corrosion resistant. Raw material is heat code traceable.
These cylinders comply with DOT-3E 1800 (US), which states it can contain flammable liquids or gases during transport. Not exceeding the pressure rating of the vessel. These cylinders also comply with TPED overseas (Europe).
Order two SS valves, Z146064, Z146056, or Z168637 separately. DOT 3E-1800.
Choose from one of the most recent versions:
Certificates of Analysis (COA)