Key Documents
Z173584
Sigma-Aldrich
Bonnet-vent tube connector
for tubing o.d., 1/8 in.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
316 stainless steel
feature
for tubing o.d. 1/8 in.
fitting
1/8 in. male NPT
General description
Use to attach vent tube to gas regulators with bonnet-vent fittings.
Choose from one of the most recent versions:
Certificates of Analysis (COA)