MilliporeSigma
All Photos(9)
Key Documents
Z173290
Sigma-Aldrich
Aldrich® Schlenk-type flasks and tubes
50ml, S/T14/20M joint
About This Item
UNSPSC Code:
41100000
material
borosilicate glass
round bottom
sterility
non-sterile
feature
closure type Screw cap
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
50 mL
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)