Key Documents
Z172960
Sigma-Aldrich
Aldrich® pour-out adapter
male joint: ST/NS 19/22
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
male joint: ST/NS19/22
General description
For pouring from flasks or funnels without pouring over ground joint.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)