Key Documents
Z171867
Sigma-Aldrich
Metal balls for bench-top laboratory roller mill
steel, diam. 1/4 in.
About This Item
UNSPSC Code:
41101708
NACRES:
NB.74
material
steel
diam.
1/4 in.
Other Notes
Uniformly through-hardened to 62-65 Rockwell C; excellent wearing qualities.
Choose from one of the most recent versions:
Certificates of Analysis (COA)