Key Documents
Z171859
Sigma-Aldrich
Bench-top laboratory roller mill
portable, capacity 2 qt , 6 lb
About This Item
UNSPSC Code:
41121800
NACRES:
NB.74
feature
portable
availability
not available in EU
capacity
2 qt , 6 lb
General description
Use with any standard pint, or quart can (unlined metal cans) for blending dry powders or with the grinding media (Z171867) listed below for grinding pigment/liquid compositions. Has heavy, continuous-duty, fan-cooled motor with overload protection. 115 V.
Choose from one of the most recent versions:
Certificates of Analysis (COA)