Key Documents
Z171778
Sigma-Aldrich
Jiffy mixer
blade diam. 2 5/8 in.,
no. of blades: 1
Synonym(s):
jiffy mixers, lab mixer, lab mixers
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
material
stainless steel
description
for container cap. 1 - 2 gal
blade diam.
2 5/8 in.
shaft diam. × L
1/4 in. × 15 in.
General description
One-piece, For general-purpose mixing using motor-driven stirrers. Aids in speedy, thorough mixing yet is nonsplashing and does not draw air. Cleans easily and quickly.
Choose from one of the most recent versions:
Certificates of Analysis (COA)