Key Documents
Z171107
Sigma-Aldrich
VICI Pressure--Lok® C-120 liquid microsyringe
capacity 10 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle
needle size
2 in.
feature
needle bevel tip
manufacturer/tradename
VICI 120022
capacity
10 μL
needle L
2 in.
Features and Benefits
In addition to standard Pressure-Lok features, has a barrel extension beyond rear flange that permits attachment of a vacuum bulb or line for easier filling or cleaning. Fixed needle.
Legal Information
Pressure-Lok is a registered trademark of Dynatech Precision Sampling Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)