Key Documents
Z170933
Supelco
VICI Pressure--Lok® C-160 liquid microsyringe
Removable needle, capacity 100 μL
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
bevel tip stainless steel needle
needle size
2.25 in.
feature
needle bevel tip
manufacturer/tradename
VICI 160225
capacity
100 μL
needle L
2.25 in.
General description
General-purpose GC syringe.
Features and Benefits
Each syringe is hand-calibrated for consistent delivery. The leaktight PTFE plunger tip eliminates freeze-up or residue build-up and allows use with gases (up to 250psi) as well as liquids. Has a positive plunger stop to prevent damage to the plunger. Barrel made of borosilicate glass. Has PTFE seal at rear of needle to isolate sample from needle cement and contamination. Shaped rear flange provides secure finger grip. Syringes with removable needles have a pressure tight, low dead-volume needle connection for a leaktight fit.
Legal Information
Pressure-Lok is a registered trademark of Dynatech Precision Sampling Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)