Key Documents
Z170410
Lightnin® 316SS mixer shaft
Model L1U08, G3Y05R
About This Item
UNSPSC Code:
41103800
NACRES:
NB.72
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Model L1U08, G3Y05R
Lightnin 143727-316
shaft diam. × L
3/8 in. × 24 in.
General description
Shaft stepped to 5/16 in. at impeller end.
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Choose from one of the most recent versions:
Certificates of Analysis (COA)