Key Documents
Z170380
Lightnin® 316 stainless steel impeller
Dispersing impeller, Model R-500
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
manufacturer/tradename
Model R-500
Lightnin 869580
bore size
5/16 in.
diam.
3.0 in. , overall
General description
Provides low flow with highest shear, excellent for solids or liquid dispersions.
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Choose from one of the most recent versions:
Certificates of Analysis (COA)