Key Documents
Z170313
Lightnin® 316 stainless steel impeller
Propeller, Model A-100, diam. 1.0 in. , overall
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
manufacturer/tradename
Model A-100
Lightnin 800570
bore size
5/16 in.
diam.
1.0 in. , overall
General description
Superpitch design with 1.5 pitch ratio for efficient mixing of hard to wet powders, standard axial flow
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Choose from one of the most recent versions:
Certificates of Analysis (COA)