Key Documents
Z170291
Lightnin® 316 stainless steel impeller
Impeller, Model R-100, diam. 1 1/2 in. , overall
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
manufacturer/tradename
Model R-100
Lightnin 809414
bore size
5/16 in.
diam.
1 1/2 in. , overall
General description
Provides high shear with moderate pumping action.
Legal Information
Lightnin is a registered trademark of Lightnin, a unit of General Signal
Choose from one of the most recent versions:
Certificates of Analysis (COA)