Key Documents
Z170011
Scienceware® glove box adapter
Threaded metal
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
label
D
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H50025-0010
General description
Tube, 21/2 in. L with 3/8 in./27 thread along complete length. Inserted in a 3/8 in. threaded hole drilled anywhere in glove box, gives threaded tube on inside and outside of box for attaching needle valves or hose ends. Complete with nuts, gaskets, and washers to seal hole.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)