Key Documents
Z169080
Sigma-Aldrich
Petri-dish rack
Overall L × W × H 4 1/2 in. × 9 in. × 10 1/2 in., pkg of 1 ea
About This Item
UNSPSC Code:
41122800
NACRES:
NB.35
material
polystyrene
packaging
pkg of 1 ea
manufacturer/tradename
Sigma-Aldrich
Overall L × W × H
4 1/2 in. × 9 in. × 10 1/2 in.
General description
Clear plastic, two-compartment rack with handle for stacking, storing, and transporting up to 26 standard petri dishes. Curved walls prevent sliding. Can be refrigerated and incubated (up to 40°C); cannot be autoclaved. 4 1/2 × 9 × 10 1/2 in.
Choose from one of the most recent versions:
Certificates of Analysis (COA)