Key Documents
Z168580
Thermolyne® Cimarec®-top stirring hotplate
AC/DC input 240 V AC, plate size 7 in. × 7 in.
About This Item
UNSPSC Code:
41102406
sterility
non-sterile
feature
CSA approved
manufacturer/tradename
Barnstead International SP46920-33
plate size
7 in. × 7 in.
size
, no of position=5
output
1100 watts
General description
Has all features of the Cimarec-top hot plate plus has excellent low-speed stirring control and speed range of 60 to 1,000 rpm. All units are CSA listed.
Legal Information
Cimarec is a registered trademark of Barnstead/Thermolyne Corp.
Thermolyne is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)