Recommended Products
description
range mg/L 0, 10, 25, 50, 100, 250, 500
semi-quantitative
specific analyte(s)
sulfite
packaging
pack of 100 ea
manufacturer/tradename
Macherey-Nagel
color
white to salmon change
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
- Fast; dip and read
- No apparatus needed, only single test stick for analysis
- Highly specific; interferences can be eliminated by precipitation or masking
- Dim.: 6 × 95 × 0.2 mm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service