Recommended Products
description
range mg/L 0, 2, 5, 10, 25, 50, 100
semi-quantitative
specific analyte(s)
iron
packaging
pack of 100 ea
manufacturer/tradename
Macherey-Nagel
color
white to blue-violet change
application(s)
environmental
detection method
colorimetric
Looking for similar products? Visit Product Comparison Guide
General description
- Fast; dip and read
- No apparatus needed, only single test stick for analysis
- Highly specific; interferences can be eliminated by precipitation or masking
- Dim.: 6 × 95 × 0.2 mm
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service