Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z164410

Twistit four-in-one rubber stoppers

set of 33 assorted sizes

Synonym(s):

rubber plug, rubber plugs, rubber stopper

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

General description

Contains 9 of size 3, 8 of size 4, 6 of size 5, 4 of size 6, 2 of size 7, 2 of size 8, 1 of size 9, and 1 of size 10.
Twist-off seals provide multiple uses of one stopper. Made of soft, white, natural rubber having high resistance to deformation after autoclaving. Has lead countersunk into holes and is vacuum tested at 30 mm Hg.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service