Key Documents
Z164364
Sigma-Aldrich
Twistit four-in-one rubber stoppers
size 6
Synonym(s):
rubber plug, rubber plugs, rubber stopper
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
size
6
size × L
32 mm × 26 mm × 25 mm
General description
Twist-off seals provide multiple uses of one stopper. Made of soft, white, natural rubber having high resistance to deformation after autoclaving. Has lead countersunk into holes and is vacuum tested at 30 mm Hg.
Choose from one of the most recent versions:
Certificates of Analysis (COA)