Z163597
MiniPax® absorbent packets
indicating silica gel, weight 10 g (desiccant), L × W 1 1/2 in. × 2 3/4 in.
About This Item
Recommended Products
manufacturer/tradename
Multisorb 02-00040AG91
L × W
1 1/2 in. × 2 3/4 in.
weight
10 g (desiccant)
Looking for similar products? Visit Product Comparison Guide
General description
Envelopes are formed from sheet of polyethylene fibers has high wet strength and is permeable to both gases and odors. Indicating MiniPax® contain cobalt chloride and should not be used for food or pharmaceutical applications.
- Compact - The low profile and compact shape of MiniPax® packets saves space and displaces a minimum amount of product. Its flat contour eliminates any potential confusion with a capsule or a pill
- Maximum surface exposure - The exceptionally narrow seal skirt on only two ends gives MiniPax® packets an unusually large adsorptive surface area for its overall size
- Integral window - An integral transparent strip permits observation of the contained adsorbent and allows effective use of indicating desiccants to show when moisture has been adsorbed and desiccant should be replaced
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 3 - Carc. 1B
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service